Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F-1 Rabbit anti-Human, Clone: JJ092-02, Novus Biologicals™
SDP

Catalog No. NBP267899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267899 Supplier Novus Biologicals Supplier No. NBP267899
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

E2F-1 Monoclonal specifically detects E2F-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen E2F-1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone JJ092-02
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias E2F transcription factor 1, E2F-1, PBR3, PRB-binding protein E2F-1, RBAP1, RBAP-1, RBBP3, RBBP-3, RBP3, Retinoblastoma-associated protein 1, Retinoblastoma-binding protein 3, transcription factor E2F1
Gene Symbols E2F1
Host Species Rabbit
Immunogen Synthetic peptide within Human E2F-1 aa 371-404/ 437. (SwissProt: Q01094 Human; SwissProt: Q61501 Mouse; SwissProt: O09139 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1869
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.