Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F6 Antibody (PCRP-E2F6-1F8), FITC, Novus Biologicals™
SDP

Catalog No. NB389102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389102 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389102 Supplier Novus Biologicals Supplier No. NBP308952F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

E2F6 Monoclonal antibody specifically detects E2F6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen E2F6
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-E2F6-1F8
Conjugate FITC
Formulation PBS
Gene Alias E2F transcription factor 6, E2F-6MGC111545, transcription factor E2F6
Host Species Mouse
Immunogen Recombinant full-length human E2F6 protein (Uniprot: O75461)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1876
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.