Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F7 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB291208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291208 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291208 Supplier Novus Biologicals Supplier No. NBP152648PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E2F7 Polyclonal antibody specifically detects E2F7 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen E2F7
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias E2F transcription factor 7, E2F-7, FLJ12981, transcription factor E2F7
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human E2F Transcription Factor 7 using the numbering given in entry NP_976328.2 (GeneID 144455).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 144455
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.