Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7204845 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8734 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8734 Supplier Proteintech Group Inc Supplier No. 134251AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E2F transcription factors, such as E2F8, are essential for orchestrating expression of genes required for cell cycle progression and proliferation.

Specifications

Antigen E2F8
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene E2F8
Gene Accession No. A0AVK6, F1LMN3, Q58FA4
Gene Alias E2F 8, E2F transcription factor 8, E2F8, Transcription factor E2F8
Gene Symbols E2f8
Host Species Rabbit
Immunogen E2F8 Fusion Protein Ag4216
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108961, 308607, 79733
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.