Learn More
R&D Systems™ E2Select Ubiquitin Conjugation Kit
Catalog No. k982
Click to view available options
Quantity:
1 kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Rapid and economical method to identify E2 enzyme(s) that functionally interact with an E3 ubiquitin ligase in vitro
  • A rapid and economical method to identify E2 enzyme(s) that functionally interact with an E3 ubiquitin ligase in vitro
  • Eliminates the need to purchase multiple enzymes individually when attempting to identify which E2 enzyme is involved in a particular ubiquitination reaction
Specifications
Includes
  • Each kit contains reagents sufficient to perform 2 ubiquitination reactions for each E2 enzyme included in the kit.
  • 96-well plate (contains a panel of 26 E2 conjugating enzymes arrayed in duplicate)
  • 10X Reaction Buffer
  • 10X E1 Enzyme
  • 20X Ubiquitin
  • 4X Mg2+-ATP
  • 5X SDS-PAGE Buffer
  • Foil Plate Sealer
Content And Storage Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Type E2Select Ubiquitin Conjugation Kit
Quantity 1 kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.