Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E4F1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB292244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292244 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292244 Supplier Novus Biologicals Supplier No. NB1002871F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E4F1 Polyclonal specifically detects E4F1 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen E4F1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias E4F transcription factor 1p50E4F, E4FMGC99614, EC 6.3.2.-, p120E4F, Putative E3 ubiquitin-protein ligase E4F1, Transcription factor E4F, transcription factor E4F1
Host Species Rabbit
Immunogen Maps to a region between residue 725 and the C-terminus (residue 784) of human E4F1 (p120E4F) using the numbering given in entry NP_004415.1 (GeneID 1877).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1877
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.