Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E6AP/UBE3A Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB50240A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50240A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50240A532 Supplier Novus Biologicals Supplier No. NB500240AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E6AP/UBE3A Polyclonal specifically detects E6AP/UBE3A in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen E6AP/UBE3A
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A
Gene Symbols UBE3A
Host Species Rabbit
Immunogen A synthetic peptide mapped to a region between residue 400 and 450 of human Ubiquitin Protein Ligase E3A using the numbering given in entry NP_000453.2 (GeneID 7337).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7337.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.