Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E6AP/UBE3A Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB318056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318056 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318056 Supplier Novus Biologicals Supplier No. NBP298953PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

E6AP/UBE3A Polyclonal antibody specifically detects E6AP/UBE3A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen E6AP/UBE3A
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ANCREPVE6AP, AS, CTCL tumor antigen se37-2, E6AP, E6-AP, E6AP ubiquitin-protein ligase, EC 6.3.2.-, FLJ26981, HPVE6A, human papilloma virus E6-associated protein, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54, ubiquitin protein ligase E3A, ubiquitin-protein ligase E3A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human E6AP/UBE3A.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7337
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.