Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT1/GLAST-1/SLC1A3 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N101869A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N101869A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N101869A750 Supplier Novus Biologicals Supplier No. NB1001869AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EAAT1/GLAST-1/SLC1A3 Polyclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen EAAT1/GLAST-1/SLC1A3
Applications ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3
Gene Symbols SLC1A3
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [Uniprot: P24942]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6507
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.