Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT2/GLT1 Antibody (28F9), Novus Biologicals™
SDP

Catalog No. NB298333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298333 100 μL
NB298332 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298333 Supplier Novus Biologicals Supplier No. NBP326322100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen EAAT2/GLT1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 28F9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human EAAT2/GLT1 [UniProt P43004]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Hypoxia, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6506
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.