Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT2/GLT1 Antibody (28F9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | EAAT2/GLT1 |
---|---|
Clone | 28F9 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
EAAT2/GLT1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
A synthesized peptide derived from Human EAAT2/GLT1 [UniProt P43004] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
28F9 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Hypoxia, Neuroscience, Neurotransmission | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6506 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title