Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT2/GLT1 Antibody - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP120136SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120136SS 0.025 mL
NBP120136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP120136SS Supplier Novus Biologicals Supplier No. NBP120136SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

EAAT2/GLT1 Polyclonal specifically detects EAAT2/GLT1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In-situ Hybridization, In vivo assay.

Specifications

Antigen EAAT2/GLT1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000, In-situ Hybridization, In vivo assay
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P43006
Gene Alias GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2
Gene Symbols SLC1A2
Host Species Rabbit
Immunogen A synthetic peptide from mouse EAAT2/GLT1 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and human.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Hypoxia, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6506
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.