Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT2/GLT1 Antibody - Azide Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$243.50 - $584.50
Specifications
Antigen | EAAT2/GLT1 |
---|---|
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000, In-situ Hybridization, In vivo assay |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP120136SS
![]() |
Novus Biologicals
NBP120136SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP120136
![]() |
Novus Biologicals
NBP120136 |
0.1 mL |
Each for $584.50
|
|
|||||
Description
EAAT2/GLT1 Polyclonal specifically detects EAAT2/GLT1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In-situ Hybridization, In vivo assay.Specifications
EAAT2/GLT1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Monkey | |
P43006 | |
6506 | |
A synthetic peptide from mouse EAAT2/GLT1 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and human. | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. |
Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000, In-situ Hybridization, In vivo assay | |
Polyclonal | |
Rabbit | |
Hypoxia, Neuroscience | |
Lyophilized from whole antisera with No Preservative | |
GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
SLC1A2 | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title