Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT2/GLT1 Rabbit anti-Human, Mouse, Rat, Clone: 7K5F7, Novus Biologicals™
SDP

Catalog No. NB167581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB167581 Supplier Novus Biologicals Supplier No. NBP316245100UL

Rabbit Monoclonal Antibody

EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen EAAT2/GLT1
Applications Western Blot
Classification Monoclonal
Clone 7K5F7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-250 of human EAAT2/GLT1 (P43004).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Hypoxia, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6506
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.