Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT2/GLT1 Rabbit anti-Human, Mouse, Rat, Clone: 7K5F7, Novus Biologicals™

Rabbit Monoclonal Antibody
$479.00
Specifications
Antigen | EAAT2/GLT1 |
---|---|
Clone | 7K5F7 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
EAAT2/GLT1 Monoclonal antibody specifically detects EAAT2/GLT1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
EAAT2/GLT1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Hypoxia, Neuroscience, Neurotransmission | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6506 | |
IgG | |
Affinity purified |
7K5F7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
GLT1, member 2, solute carrier family 1 (glial high affinity glutamate transporter), member 2, Solute carrier family 1 member 2 | |
A synthetic peptide corresponding to a sequence within amino acids 150-250 of human EAAT2/GLT1 (P43004). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title