Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT3 Antibody (4D6.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019449 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019449 0.025 mg
NBP229747 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019449 Supplier Novus Biologicals Supplier No. NBP2297470.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EAAT3 Monoclonal specifically detects EAAT3 in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen EAAT3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4D6.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml
Formulation 20mM Phosphate (pH 7.6) and 0.25M NaCl with 0.1% Sodium Azide
Gene Accession No. P43005
Gene Alias EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1
Gene Symbols SLC1A1
Host Species Mouse
Immunogen C-terminus peptide (KDKSDTISFTQTSQF).
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6505
Test Specificity Glutamate Transporter EAAC1. Based on protein sequence the antibody is expected to react with mouse, bovine and rabbit.
Target Species Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.