Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16251920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16251920 20 μL
NBP162519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16251920 Supplier Novus Biologicals Supplier No. NBP16251920UL

Rabbit Polyclonal Antibody

EAAT3 Polyclonal specifically detects EAAT3 in Human samples. It is validated for Western Blot.

Specifications

Antigen EAAT3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P43005
Gene Alias EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1
Gene Symbols SLC1A1
Host Species Rabbit
Immunogen The immunogen for anti-SLC1A1 antibody: synthetic peptide directed towards the N terminal of human SLC1A1 (NP_004161). Peptide sequence VLVREHSNLSTLEKFYFAFPGEILMRMLKLIILPLIISSMITGVAALDSN.
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6505
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.