Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAAT3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | EAAT3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15988020
![]() |
Novus Biologicals
NBP15988020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159880
![]() |
Novus Biologicals
NBP159880 |
100 μL |
Each for $487.50
|
|
|||||
Description
EAAT3 Polyclonal specifically detects EAAT3 in Human samples. It is validated for Western Blot.Specifications
| EAAT3 | |
| Polyclonal | |
| Rabbit | |
| P43005 | |
| 6505 | |
| IgG | |
| 57 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 | |
| SLC1A1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title