Learn More
EAF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 137871AP150UL

Description
13787-1-AP is a rabbit polyclonal antibody raised against the full-length human EAF1 protein.
Acts as a transcriptional transactivator of ELL and ELL2 elongation activities.Specifications
EAF1 | |
Polyclonal | |
Unconjugated | |
EAF1 | |
EAF1, ELL associated factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
306261, 74427, 85403 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96JC9, Q9D4C5 | |
EAF1 | |
EAF1 Fusion Protein Ag4804 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.