Learn More
EAF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | EAF1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9006
![]()
|
Proteintech Group Inc
137871AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
13787-1-AP is a rabbit polyclonal antibody raised against the full-length human EAF1 protein.
Acts as a transcriptional transactivator of ELL and ELL2 elongation activities.Specifications
| EAF1 | |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q96JC9, Q9D4C5 | |
| 306261, 74427, 85403 | |
| EAF1 Fusion Protein Ag4804 | |
| Primary | |
| -20°C | |
| EAF1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EAF1, ELL associated factor 1 | |
| EAF1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.