Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAR2/NR2F6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224510SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224510SS 0.025 mg
NBP224510 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224510SS Supplier Novus Biologicals Supplier No. NBP224510SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EAR2/NR2F6 Polyclonal specifically detects EAR2/NR2F6 in Human, Canine, Primate samples. It is validated for Western Blot.

Specifications

Antigen EAR2/NR2F6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias EAR-2ERBA-related gene-2, EAR2nuclear receptor subfamily 2 group F member 6, ERBAL2v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2, nuclear receptor subfamily 2, group F, member 6, V-erbA-related protein 2
Gene Symbols NR2F6
Host Species Rabbit
Immunogen A portion of amino acids 20-70 of human NR2F6 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2063
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, 92% homologous indog, 85% homologous incow and opossum and 78% homologous in mouse, horseand rat.
Target Species Human, Canine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.