Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EARS2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156400
Description
EARS2 Polyclonal specifically detects EARS2 in Human samples. It is validated for Western Blot.Specifications
EARS2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 6.1.1.17, GluRS, Glutamate--tRNA ligase, glutamyl-tRNA synthetase 2, mitochondrial (putative), KIAA1970glutamate tRNA ligase 2, mitochondrial, MSE1, probable glutamyl-tRNA synthetase, mitochondrial | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Bovine: 92%; Guinea pig: 92%; Mouse: 92%; Rat: 92%; Chicken: 85%; Canine: 85%; Equine: 85%; Rabbit: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q86YH3 | |
EARS2 | |
Synthetic peptides corresponding to EARS2(glutamyl-tRNA synthetase 2, mitochondrial (putative)) The peptide sequence was selected from the middle region of EARS2. Peptide sequence TAKHLLLYQALGWQPPHFAHLPLLLNRDGSKLSKRQGDVFLEHFAADGFL. | |
100 μL | |
Lipid and Metabolism | |
124454 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction