Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EARS2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096248 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096248 20 μL
NB096247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096248 Supplier Novus Biologicals Supplier No. NBP30502720UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EARS2 Polyclonal specifically detects EARS2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen EARS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 6.1.1.17, GluRS, Glutamate--tRNA ligase, glutamyl-tRNA synthetase 2, mitochondrial (putative), KIAA1970glutamate tRNA ligase 2, mitochondrial, MSE1, probable glutamyl-tRNA synthetase, mitochondrial
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 42-220 of human EARS2 (NP_001295140.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 124454
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.