Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters

Catalog No. p-6024224
Click to view available options
Type:
Capillary EASY-Spray Emitter w/ Transfer Line
Capillary EASY-Spray Emitter, Bullet Type w/o Transfer Line
Nano EASY-Spray Emitter w/ Transfer Line
Nano EASY-Spray Emitter, Bullet Type w/o Transfer Line
Diameter (Metric):
0.02 mm
0.075 mm
Emitter: 10 μm I.D.
Emitter: 15 μm I.D.

Use this easy-to-use sprayer in your sensitive LC-MS separation of protein and peptides using nano and capillary LC.

Connect your nano or capillary column to your Thermo Scientific™ mass spectrometers (MS) more easily using Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters. The nano and capillary emitters act as a column-independent sprayer, allowing the introduction of low flow from nano and capillary columns without troublesome handling of traditional spray needles. There are two versions of emitters available: with and without integrated transfer lines. These sprayers are ideal for protein and peptide separation using nano and capillary HPLC.

For sensitive protein and peptide separation using nano and capillary HPLC
These emitters offer easy connection to Thermo Scientific™ MAbPac™ and Accucore™ nano and capillary columns for enhanced separation and MS detection of proteins, peptides and mnoclonal antibodies (mAbs).

TRUSTED_SUSTAINABILITY

Specifications

Column Format EASY-Spray Emitter
Diameter (Metric) 0.02 mm
Includes Transfer line: 20 μm I.D. x 50 cm Length
Length (Metric) 500 mm
Product Line Easy Spray
Packing Material Unpacked
Column Type Emitter
Type Nano EASY-Spray Emitter w/ Transfer Line

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.