Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBAG9/RCAS1 Antibody (019), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB038374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038374 Supplier Novus Biologicals Supplier No. NBP290266AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EBAG9/RCAS1 Monoclonal specifically detects EBAG9/RCAS1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen EBAG9/RCAS1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 19
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EBAG9/RCAS1 (O00559; Arg28-Ser213).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9166
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.