Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBF-3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17998220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17998220 20 μL
NBP179982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17998220 Supplier Novus Biologicals Supplier No. NBP17998220UL

Rabbit Polyclonal Antibody

EBF-3 Polyclonal specifically detects EBF-3 in Human samples. It is validated for Western Blot.

Specifications

Antigen EBF-3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001005463
Gene Alias COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF-3, Olf-1/EBF-like 2, transcription factor COE3
Gene Symbols EBF3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human EBF3. Peptide sequence AHTGMMGVNSFSSQLAVNVSETSQANDQVGYSRNTSSVSPRGYVPSSTPQ.
Molecular Weight of Antigen 60 kDa
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253738
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.