Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBF-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | EBF-3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17998220
![]() |
Novus Biologicals
NBP17998220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179982
![]() |
Novus Biologicals
NBP179982 |
100 μL |
Each for $487.50
|
|
|||||
Description
EBF-3 Polyclonal specifically detects EBF-3 in Human samples. It is validated for Western Blot.Specifications
EBF-3 | |
Polyclonal | |
Purified | |
RUO | |
COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF-3, Olf-1/EBF-like 2, transcription factor COE3 | |
EBF3 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
NP_001005463 | |
253738 | |
Synthetic peptide directed towards the middle region of human EBF3. Peptide sequence AHTGMMGVNSFSSQLAVNVSETSQANDQVGYSRNTSSVSPRGYVPSSTPQ. | |
Primary | |
60 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title