Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBF-3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216254 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216254 Supplier Novus Biologicals Supplier No. NBP216254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EBF-3 Polyclonal specifically detects EBF-3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen EBF-3
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.34mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9H4W6
Gene Alias COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF-3, Olf-1/EBF-like 2, transcription factor COE3
Gene Symbols EBF3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EBF3. The exact sequence is proprietary.
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253738
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (99%), Rat (99%), Xenopus laevis (95%), Zebrafish (91%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.