Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI2/GPR183 Antibody (486633) [Biotin], Novus Biologicals™
SDP

Catalog No. NB402100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402100 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402100 Supplier Novus Biologicals Supplier No. FAB10272B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBI2/GPR183 Monoclonal antibody specifically detects EBI2/GPR183 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen EBI2/GPR183
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 486633
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor), EBV-induced G-protein coupled receptor 2, Epstein-Barr virus-induced G-protein coupled receptor 2, G protein-coupled receptor 183, G-protein coupled receptor 183
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived transfected withrecombinant human EBI2, Accession # P32249
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1880
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.