Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Antibody (10J803), PerCP, Novus Biologicals™
SDP

Catalog No. NBP203940P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203940P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP203940P Supplier Novus Biologicals Supplier No. NBP203940PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EBI3 Monoclonal specifically detects EBI3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Classification Monoclonal
Clone 10J803
Conjugate PerCP
Dilution Flow Cytometry 1 μL/1 million cells
Gene Alias cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta
Gene Symbols EBI3
Host Species Rat
Immunogen Full-length recombinant mouse EBI3 protein
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10148
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.