Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Antibody (15k8D10) - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP2273635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2273635 0.5 mg
NBP227363 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2273635 Supplier Novus Biologicals Supplier No. NBP2273630.5MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 15k8D10
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Formulation Sterile - filtered PBS with No Preservative
Gene Alias cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta
Gene Symbols EBI3
Host Species Mouse
Immunogen A full-length human Interleukin-35-Fc fusion protein was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10148
Test Specificity The human Interleukin-35 protein is a dimeric protein consisting of the human IL-12a/p35 subunit (NP_000873.2) and the human EBI3 (NP_005746) protein. This clone has been tested to be specific to the EBI3 subunit of the target protein. Clone 15K8D10 tests as being specific to the EBI3 subunit.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.