Learn More
Description
Specifications
Specifications
| Antigen | IL-27/IL-35 EBI3 Subunit |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 5P10D3 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry |
| Gene Alias | cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta |
| Gene Symbols | EBI3 |
| Host Species | Rat |
| Immunogen | Full-length recombinant mouse EBI3 protein |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
