Learn More
Description
Specifications
Specifications
| Antigen | IL-27/IL-35 EBI3 Subunit |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 5P10D3 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-5 ug/ml, Flow Cytometry 1-3ug/10^6cells, ELISA reported in scientific literature (PMID 30660991) |
| Gene Accession No. | O35228 |
| Gene Alias | cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta |
| Gene Symbols | EBI3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
