Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBNA1 Antibody (E1-2.5), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB500312G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500312G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500312G Supplier Novus Biologicals Supplier No. NB500312G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBNA1 Monoclonal specifically detects EBNA1 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EBNA1
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone E1-2.5
Conjugate DyLight 488
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias BKRF1, Epstein-Barr nuclear antigen 1, HHV4gp098, nuclear antigen EBNA1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the repetitive Gly-Ala region of EBNA-1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody E1-2.5 recognizes repetitive Gly-Ala region of the Epstein-Barr virus (EBV) nuclear antigen 1 (EBNA-1). EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.