Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBNA1BP2/EBNA1 Binding Protein 2/EBP2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | EBNA1BP2/EBNA1 Binding Protein 2/EBP2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9443
![]() |
Bethyl Laboratories, Inc
A305067AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
EBNA1BP2 is required for the processing of the 27S pre-rRNA.Specifications
EBNA1BP2/EBNA1 Binding Protein 2/EBP2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
EBNA1-binding protein 2, EBP2, NOBP, nuclear FGF3 binding protein, nucleolar protein p40, nucleolar protein p40; homolog of yeast EBNA1-binding protein, P40 | |
EBNA1BP2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q99848 | |
10969 | |
Between 256 and 306 | |
Primary | |
4° C | |
EBNA1BP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title