Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBOV GP Antibody (02), PE, Novus Biologicals™
SDP

Catalog No. NB327507 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327507 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327507 Supplier Novus Biologicals Supplier No. NBP305998PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBOV GP Monoclonal antibody specifically detects EBOV GP in Virus samples. It is validated for ELISA

Specifications

Antigen EBOV GP
Applications ELISA
Classification Monoclonal
Clone 02
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV GP (Accession#: AHX24649.1; Met1-Gln650).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3160774
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.