Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBOV GP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB142596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB142596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB142596 Supplier Novus Biologicals Supplier No. NBP305820AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EBOV GP1 Polyclonal specifically detects EBOV GP1 in Ebola Virus samples. It is validated for Western Blot.

Specifications

Antigen EBOV GP1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) GP1/ Glycoprotein (ACI28624.1; Met1-Arg501).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3160774
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.