Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBOV Nucleoprotein Antibody (09), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB388795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388795 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388795 Supplier Novus Biologicals Supplier No. NBP312759AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBOV Nucleoprotein Monoclonal antibody specifically detects EBOV Nucleoprotein in Ebola virus samples. It is validated for Western Blot

Specifications

Antigen EBOV Nucleoprotein
Applications Western Blot
Classification Monoclonal
Clone 09
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV Nucleoprotein (Accession#: AHX24646.1; His630-Gln739).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Ebola virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.