Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBP Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB133828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133828 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133828 Supplier Novus Biologicals Supplier No. NBP306227AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EBP Polyclonal specifically detects EBP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen EBP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 3-beta-hydroxysteroid-Delta(8), CDPX2, Cholestenol Delta-isomerase, CPX3-beta-hydroxysteroid-delta-8, CPXDX-linked dominant (Happle syndrome), D8-D7 sterol isomerase, Delta(7)-isomerase, Delta(8)-Delta(7) sterol isomerase, delta-7-isomerase, EC 5.3.3.5, emopamil binding protein (sterol isomerase), Emopamil-binding protein, sterol 8-isomerase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EBP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10682
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.