Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBV Early Antigens Antibody (1108-1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP234516FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234516FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234516FR Supplier Novus Biologicals Supplier No. NBP234516FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBV Early Antigens Monoclonal specifically detects EBV Early Antigens in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EBV Early Antigens
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1108-1
Conjugate DyLight 680
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Gene Alias Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like
Host Species Mouse
Immunogen Affinity purified early antigen polypeptides from induced Raji cells precipitated with African Burkitt's lymphoma sera
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Epstein-Barr virus (EBV), also designated human herpesvirus 4 (HHV-4), is a member of the herpesvirus family and is one of the most common human viruses. EBV infects B cells and, though often asymptomatic, it can cause infectious mononucleosis, a disease characterized by fatigue, fever, sore throat and muscle soreness. The EBV-induced early antigens (Ea) are among several antigen complexes that have been identified in EBV-infected cells. The Ea complex is composed of diffuse (Ea-D) and restricted (Ea-R) components. The activity of Ea-D is suppressed during latent infection. BMRF1, the gene that encodes for Ea-D, is closely associated with the gene encoding for EBV DNA polymerase, and Ea-D is essential for the activity of this polymerase. Ea-D forms a complex with EBV DNase and, together, they may play a role in viral replication.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.