Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBV Latent Membrane Protein 1 Antibody (LMPO24), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250383AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250383AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250383AI Supplier Novus Biologicals Supplier No. NBP250383AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EBV Latent Membrane Protein 1 Monoclonal specifically detects EBV Latent Membrane Protein 1 in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Intracellular).

Specifications

Antigen EBV Latent Membrane Protein 1
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone LMPO24
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunohistochemistry, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Purified plasma membranes from EBV Latent Membrane Protein 1 expressing insect cells infected with a recombinant EBV Latent Membrane Protein 1 baculovirus
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Niedobitek et al., 1995 (PMID: 7605996): “The LMPO24 clone demonstrates strong and specific reactivity in immunohistochemical staining of paraffin-embedded tissues”.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.