Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECE-1 Antibody (303913), PerCP, Novus Biologicals™
SDP

Catalog No. NB367679 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367679 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367679 Supplier Novus Biologicals Supplier No. FAB17841C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ECE-1 Monoclonal specifically detects ECE-1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen ECE-1
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 303913
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EC 3.4.24, EC 3.4.24.71, ECE, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ECE-1, Gln90-Trp770, Accession # P42892
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1889
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.