Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECE-1 Antibody (OTI2B6), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270612B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270612B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270612B Supplier Novus Biologicals Supplier No. NBP270612B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ECE-1 Monoclonal specifically detects ECE-1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ECE-1
Applications Western Blot
Classification Monoclonal
Clone OTI2B6
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.4.24, EC 3.4.24.71, ECE, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1
Gene Symbols ECE1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 486-770 of human ECE-1 (NP_001388) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1889.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.