Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECHS1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB041524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041524 Supplier Novus Biologicals Supplier No. NBP296977B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECHS1 Polyclonal specifically detects ECHS1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ECHS1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias EC 4.2.1, enoyl CoA hydratase, short chain, 1, mitochondrial, enoyl Coenzyme A hydratase, short chain, 1, mitochondrial, Enoyl-CoA hydratase 1, enoyl-CoA hydratase, mitochondrial, SCEHEC 4.2.1.17, Short-chain enoyl-CoA hydratase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ECHS1 (AAH08906.1; 28A-290Q).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1892
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.