Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECM-1/Secretory Component P85 Antibody (001), HRP
SDP

Catalog No. NB074737 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074737 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074737 Supplier Novus Biologicals Supplier No. NBP290539H

Rabbit Monoclonal Antibody

ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Mouse samples. It is validated for ELISA.

Specifications

Antigen ECM-1/Secretory Component P85
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate HRP
Formulation PBS
Gene Alias ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ECM-1/Secretory Component P85 (Q61508-1; Met 1-Glu 559).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1893
Test Specificity Has cross-reactivity in ELISA with: Human ECM1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.