Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECM-1/Secretory Component P85 Antibody (ECM1/2889R), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB386826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386826 Supplier Novus Biologicals Supplier No. NBP308458DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ECM-1/Secretory Component P85 Monoclonal antibody specifically detects ECM-1/Secretory Component P85 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ECM-1/Secretory Component P85
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ECM1/2889R
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85
Host Species Rabbit
Immunogen Recombinant human full-length ECM-1/Secretory Component P85 protein (Uniprot: Q16610)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Extracellular Matrix, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1893
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.