Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECM-1/Secretory Component P85 Antibody (ECM1/792) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247804A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247804A 0.1 mg
NBP247804B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247804A Supplier Novus Biologicals Supplier No. NBP2478040.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ECM-1/Secretory Component P85 Monoclonal specifically detects ECM-1/Secretory Component P85 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.

Specifications

Antigen ECM-1/Secretory Component P85
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Classification Monoclonal
Clone ECM1/792
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85
Gene Symbols ECM1
Host Species Mouse
Immunogen Recombinant human ECM1 protein
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Extracellular Matrix, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1893
Test Specificity This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiation and tumorigenesis, all processes associated with differential secretory component expression.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.