Learn More
ECM-1/Secretory Component P85 Rabbit anti-Human, Clone: 30, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | ECM-1/Secretory Component P85 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 30 |
| Conjugate | Unconjugated |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human ECM-1/Secretory Component P85 Protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.