Learn More
United Scientific™ EcoLift Lab Jack
Click to view available options
Height (Metric):
6.35 to 15 cm
6.35 to 25.4 cm
7.6 to 30.5 cm
Description
The EcoLift™ Lab Jacks are constructed of powder coated stainless steel. The height can be easily adjusted with a simple turn of the comfortable molded knob. The jacks are available in three sizes to fit various laboratory applications.
- Powder coating provides corrosion resistance
- Stable and durable
- Height is easily adjustable
Specifications
Specifications
Material | Powder Coated Stainless Steel |
Height (Metric) | 6.35 to 15 cm |
Product Type | Lab Jack |
Cancer and Reproductive Harm