Learn More
United Scientific™ Economy Calorimeter
missing translation for 'orderingAttributeHoverText'
:
Calorimmeter
Description
The economy food calorimeter allows students to investigate the amount of energy released when food is burned.
- This involves using the burning energy of food to heat a vessel of water
- The increase in temperature of the water represents the energy given off by the burning food
- During the investigation students will learn how much energy if required to burn off some food items, and also how to read the caloric specification on food wrappers
- Determine the caloric count of snack food
- Holds the food item safely
- Includes activity guide
- Kit Contents:
- Erlenmeyer flask, 125 mL, with plastic ring
- Metal chamber
- Metal plate lid for chamber
- Cork with pin for food support
- Activity guide
Specifications
Specifications
Product Type | Calorimeter |